Tetrabutylammonium perchlorate, Supelco®
Supplier: Merck
M-Clarity™ quality level = MQ100
86893-10G
86893-50G
86893-10GEA
61.9
GBP
86893-10G
86893-50G
Tetrabutylammonium perchlorate, Supelco®
Tetrabutylammonium perchlorate
Formula:
C₁₆H₃₆ClNO₄ Melting Pt: 210 °C Density: 1,07 g/cm³ (20 °C) |
MDL Number:
MFCD00038722 CAS Number: 1923-70-2 EINECS: 217-655-5 UN: 1481 ADR: 5.1,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...