tert-Butylchlorodimethylsilane
Supplier: Molekula
|
Danger
|
Synonyms:
tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
38882574-25G
38882574-100G
MOLE38882574-25GEA
68.9
GBP
MOLE38882574-25G
MOLE38882574-100G
tert-Butylchlorodimethylsilane
tert-Butylchlorodimethylsilane
Formula:
C₆H₁₅ClSi MW: 150,72 g/mol Boiling Pt: 125 °C (1013 hPa) Melting Pt: 87…90 °C Density: 0,81 g/cm³ (20 °C) Flash Pt: 22 °C |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 EINECS: 242-042-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...