Potassium orthovanadate ≥99.9% (metals basis)

Supplier: Thermo Scientific
Danger

39502.14 39502.22
39502.14EA 220 GBP
39502.14 39502.22
Potassium orthovanadate ≥99.9% (metals basis)
Potassium orthovanadate

Purity: 99.9% CAS No.: 14293-78-8 Molecular Formula: K3VO4 Formula Weight: 232.24 Physical Form: Granular Odour: Odourless UN No.: 3285 MDL No.: MFCD00210642

Formula: K₃O₄V
MW: 232,23 g/mol
Melting Pt: 1300 °C
Storage Temperature: Ambient
MDL Number: MFCD00210642
CAS Number: 14293-78-8
EINECS: 238-220-6
UN: 3285
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR