PIPES sesquisodium salt ≥99%, high purity
Supplier: VWR Chemicals
Synonyms:
2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid sesquisodium salt, Sesquisodium piperazine-1,4-diethanesulphonate, Sodium 1,1'-(piperazine-1,4-diyl)diethanesulphonate 1-(4-(1-sulphoethyl)piperazin-1-yl)ethanesulphonate(3:2), PIPES-Na1,5, Piperazine-N,N'-bis-2-ethanesulphonic acid sesquisodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) sesquisodium salt
0169-10KGEA
3850
GBP
0169-10KG
0169-1KG
PIPES sesquisodium salt ≥99%, high purity
PIPES sesquisodium salt
Formula:
C₁₆H₃₃N₄Na₃O₁₂S₄ MW: 670,69 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066393 CAS Number: 100037-69-2 |
Specification Test Results
Chloride | ≤1% |
Heavy Metals | ≤0.0001% |
pKa (25 °C) | 6.7 – 6.9 |
Purity (Anhydrous) | ≥99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...