4-Heptylphenol ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-n-Heptylphenol

A15407.03 A15407.06 A15407.14
A15407.03EA 40.8 GBP
A15407.03 A15407.06 A15407.14
4-Heptylphenol ≥98%
4-Heptylphenol

Purity: 98+% CAS No.: 1987-50-4 Molecular Formula: C13H20O Formula Weight: 192.30 Refractive Index: 1.5070 Melting Point: 25-28° Boiling Point: 156°/9mm UN No.: 2430 MDL No.: MFCD00041751

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Boiling Pt: 156 °C (9 mmHg)
Melting Pt: 25…28 °C
Density: 0.939 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041751
CAS Number: 1987-50-4
EINECS: 217-862-0
UN: 2430
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR