3-Phenyl-3-pentanol 97%
Supplier: Thermo Scientific
B20871.03
B20871.06
B20871.03EA
0
GBP
B20871.03
B20871.06
3-Phenyl-3-pentanol 97%
3-Phenyl-3-pentanol
Purity: 97%
CAS No.: 1565-71-5
Molecular Formula: C11H16O
Formula Weight: 164.25
Refractive Index: 1.5160
Boiling Point: 98-100°/3mm
MDL No.: MFCD00021830
Formula:
C₆H₅C(OH)(CH₂CH₃)₂ MW: 164,25 g/mol Boiling Pt: 98…100 °C (3 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00021830 CAS Number: 1565-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...