Bottles, Narrow Neck, Boston Round, PET, Rixius
Supplier: RIXIUS
93100050028WC
RIXI93100050028WCEA 5470 GBP
RIXI93100050028WC
Bottles, Narrow Neck, Boston Round, PET, Rixius
Bottles Plastic Bottles
Round bottle made of PET. Can be used in combination with pump dispensers.
  • Perfect alternative to glass
  • Low dead weight, which means low transport and disposal costs
  • Recyclable
  • Almost unbreakable
  • Food grade
Particularly suitable for homeopathic and cosmetic products, aromas, fragrance oils and dietary supplements.