4-Chloro-2-methoxyphenol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Chlor-2-hydroxyanisole, 4-Chloroguaiacol

A14806.14 A14806.22
A14806.14EA 0 GBP
A14806.14 A14806.22
4-Chloro-2-methoxyphenol ≥97%
4-Chloro-2-methoxyphenol

Purity: 97% CAS No.: 16766-30-6 Molecular Formula: C7H7ClO2 Formula Weight: 158.59 Flash Point: 110°(230°F) Refractive Index: 1.5595 Odour: Phenol-like Melting Point: 16-17° Boiling Point: 241° Beilstein Reference: 1940516 MDL No.: MFCD00070774

Formula: C₇H₇ClO₂
Boiling Pt: 241 °C
Melting Pt: 16…17 °C
Density: 1,304
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070774
CAS Number: 16766-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR