Methanol ≥99.8%, PHOTREX® for spectrophotometry, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Hydroxy methane
, Methyl alcohol
, Carbinol
, Wood alcohol
9069-01EA
0
GBP
9069-03
9069-01
Methanol ≥99.8%, PHOTREX® for spectrophotometry, J.T.Baker®
Methanol
Formula:
CH₃OH MW: 32,04 g/mol Boiling Pt: 64,6 °C (1013 hPa) Melting Pt: –98 °C Density: 0,7918 g/cm³ (20 °C) Flash Pt: 11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 13,05984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...