Lead ≥99.95% (metals basis), powder -100 mesh

Supplier: Thermo Fisher Scientific
Danger

45602.A1 45602.30
45602.A1EA 115 GBP
45602.A1 45602.30
Lead ≥99.95% (metals basis), powder -100 mesh
Lead

Purity: 99.95% CAS No.: 7439-92-1 Molecular Formula: Pb Physical Form: Atomized powder Odour: Odourless UN No.: 3077 MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1749 °C (1013 hPa)
Melting Pt: 327,5 °C
Density: 11,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR