Specimen pin stubs SEM
Supplier: IBS Stock Items
AGG301
IBSAAGG301EA 0 GBP
IBSAAGG301
Specimen pin stubs SEM
Specimen Mounting Supplies
SEM specimen stubs for Leo/Cambridge, Fei/Phillips, Camscan, Tescan, Zeiss instruments.
  • Pin length: 8 mm
  • Pin diameter: 3,2 mm