Supelco® Organic Reference Standard, F.A.M.E. Mix RM-1
Supplier: Merck
O7006-1AMP
O7006-1AMPEA
81.7
GBP
O7006-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix RM-1
Standards
Organic Standards
Palmitic acid methyl ester (C16:0) 6,0% Stearic acid methyl ester (C18:0) 3,0% Oleic acid methyl ester (C18:1cis-9) 35,0% Linoleic acid methyl ester (C18:2cis-912) 50,0% Linolenic acid methyl ester (C18:3cis-91215) 3,0% Arachidic acid methyl ester (C20:0) 3,0%.
Caution: P273 - P501