A real all-rounder tool for de-icing, drying, shaping, heat shrinking, paint stripping, soft-soldering and much more.
- Micanite heating element
- Three temperature settings: 50, 400 and 600 °C
- Tool can be used with all standard nozzles
- Air flow rate of 100 to 500 l/min
- Thermal cut-out
- Folding box model
The temperature not only adjusts in three stages – the airflow does too. The optimised weight balance virtually rules out fatigue while working. The ergonomically shaped handle with soft inlay ensures optimum handling and the integrated thermal cut-out prevents overheating.
Certifications: Protection class (without earth terminal) II.