Furan-2,5-diyldimethanol ≥98%, crystalline solid
Supplier: Cayman Chemical
|
Warning
|
Synonyms:
2,5-Furandimethanol, 5-(Hydroxymethyl)furfuryl Alcohol
20658-1
20658-10
20658-5
20658-500
CAYM20658-1EA
219
GBP
CAYM20658-1
CAYM20658-10
CAYM20658-5
CAYM20658-500
Furan-2,5-diyldimethanol ≥98%, crystalline solid
Furan-2,5-diyldimethanol
5-(Hydroxymethyl)furfuryl alcohol is a heterocyclic organic compound that is naturally produced by certain wood-inhabiting fungi. It can be derived by the reduction of the formyl group of 5-hydroxymethylfurfural.
5-(Hydroxymethyl)furfuryl alcohol can be used as a building block in the enzymatic synthesis of biobased polyesters.
Formula:
C₆H₈O₃ MW: 128,13 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00003253 CAS Number: 1883-75-6 |
Specification Test Results
Purity | ≥98% |
λmax | 223 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...