Methyl yellow indicator, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Dimethyl yellowSudan Yellow4-(Dimethylamino)azobenzene4-(N,N-Dimethylamino)azobenzene

33129-25GEA 66.6 GBP
33129-25G
Methyl yellow indicator, Fluka™
Methyl Yellow
Formula: C₁₄H₁₅N₃
MW: 225,29 g/mol
Melting Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00008308
CAS Number: 60-11-7
EINECS: 200-455-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Identity (IR) complying
Change complying
Extinction maximum at 403 - 413 nm
Absorbtion (1%/1cm) 1050 - 1350

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR