ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) ≥93%

Supplier: Thermo Scientific

Synonyms: NADP monosodium saltNADP-NaNicotinamide-adenine dinucleotide phosphate monosodium saltNADP SodiumNadide phosphate monosodium salt

J22655.06EA 1130 GBP
J22655.06
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) ≥93%
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
Formula: C₂₁H₂₇N₇NaO₁₇P₃
MW: 765,39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036973
CAS Number: 1184-16-3
EINECS: 214-664-6

Order Now

Specification Test Results

Assay (Enzymatic) ≥93%
Residual Water ≤8.0%
Molar Extinction Coefficient (Molar Absorptivity) epsilon₂₆₀ (pH 7.5) [18.0 ±0.8]×10³
Molar Extinction Coefficient (Molar Absorptivity) epsilon₃₄₀ (pH 7.5) [6.2 ±0.4]×10³
Spectral Ratio A₂₅₀/A₂₆₀ (pH 7.5) 0.83 ±0.03
Spectral Ratio A₂₈₀/A₂₆₀ (pH 7.5) 0.21 ±0.02
Spectral Ratio (pH 7.5, Reduced with G6PDH), A₂₆₀/A₃₄₀ 2.33 ±0.11
Spectral Ratio (pH 7.5, Reduced with G6PDH), A₃₄₀/A₂₆₀ 0.43 ±0.02
Sodium (Na) 3.0 ±1.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR