Methyl-5-amino-2-methylbenzoate 97%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 5-amino-2-methylbenzoate, 5-Amino-2-methylbenzoic acid methyll ester, 5-Amino-o-toluic acid methyll ester, Methyl 5-Amino-o-toluate

H61671.14 H61671.22
H61671.14EA 161 GBP
H61671.14 H61671.22
Methyl-5-amino-2-methylbenzoate 97%
Methyl-5-amino-2-methylbenzoate
Formula: C₉H₁₁NO₂
MW: 165,19 g/mol
Melting Pt: 39…40 °C
Storage Temperature: Freezer
MDL Number: MFCD08752568
CAS Number: 18595-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR