3-Benzoylpyridine 98,0 GC_ASSAY_METHOD zS, Sigma-Aldrich®

Supplier: Merck

Synonyms: Phenyl 3-pyridyl ketone, Phenyl(pyridin-3-yl)methanone

MQ200

8.43851.0005
8.43851.0005EA 46.2 GBP
8.43851.0005
3-Benzoylpyridine 98,0 GC_ASSAY_METHOD zS, Sigma-Aldrich®
3-Benzoylpyridine
Formula: C₁₂H₉NO
MW: 183.21 g/mol
Boiling Pt: 307
Melting Pt: 42
Storage Temperature: Ambient
MDL Number: MFCD00006394
CAS Number: 5424-19-1
EINECS: 226-561-3

Order Now

Specification Test Results

Assay (GC, area%) ≥ 98.0 % (a/a)
Melting range (lower value) ≥ 36 °C
Melting range (upper value) ≤ 40 °C
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR