2-Amino-5-chloro-3-nitropyridine ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Chloro-3-nitro-2-pyridylamine, 5-Chloro-3-nitropyridin-2-amine
B22484.14
B22484.22
B22484.14EA
97.7
GBP
B22484.14
B22484.22
2-Amino-5-chloro-3-nitropyridine ≥98%
2-Amino-5-chloro-3-nitropyridine
Purity: 98+%
CAS No.: 5409-39-2
Molecular Formula: C5H4ClN3O2
Formula Weight: 173.56
Melting Point: 190-195°
Beilstein Reference: 383850
Formula:
C₅H₄ClN₃O₂ MW: 173,56 g/mol Melting Pt: 190…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092011 CAS Number: 5409-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...