4-Chloro-3-fluoropyridine hydrochloride ≥97%
Supplier: Thermo Scientific
|
Warning
|
H27184.06
H27184.14
H27184.06EA
84.4
GBP
H27184.06
H27184.14
4-Chloro-3-fluoropyridine hydrochloride ≥97%
4-Chloro-3-fluoropyridine hydrochloride
Purity: 97%
CAS No.: 119229-74-2
Molecular Formula: C5H3ClFN.HCl
Formula Weight: 168.00
Melting Point: ca 160° subl.
MDL No.: MFCD03453233
Formula:
C₅H₄Cl₂FN Melting Pt: ∼ 160 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD03092936 CAS Number: 119229-74-2 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...