2-Chloronicotinic acid ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid

A17082.09 A17082.18 A17082.30
A17082.09EA 25.6 GBP
A17082.09 A17082.18 A17082.30
2-Chloronicotinic acid ≥99%
2-Chloronicotinic acid

Purity: 99% CAS No.: 2942-59-8 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Melting Point: ca 180° dec. Beilstein Reference: 119023

Formula: C₆H₄ClNO₂
MW: 157,56 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: ∼ 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR