3,4-Dibromo-2-fluoropyridine 95%
Supplier: Thermo Scientific
|
Danger
|
H27063.MD
H27063.03
H27063.MDEA
55.2
GBP
H27063.MD
H27063.03
3,4-Dibromo-2-fluoropyridine 95%
3,4-Dibromo-2-fluoropyridine
Purity: 95%
CAS No.: 137718-84-4
Molecular Formula: C5H2Br2FN
Formula Weight: 254.88
Melting Point: 74-77°
UN No.: 2811
MDL No.: MFCD09037460
Formula:
C₅H₂Br₂FN Melting Pt: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037460 CAS Number: 137718-84-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...