6-Bromonicotinic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid

H27926.14 H27926.06
H27926.14EA 338 GBP
H27926.14 H27926.06
6-Bromonicotinic acid ≥97%
6-Bromonicotinic acid

Purity: 97% CAS No.: 6311-35-9 Molecular Formula: C6H4BrNO2 Formula Weight: 202.01 Melting Point: 179-188° MDL No.: MFCD01927100

Formula: C₆H₄BrNO₂
MW: 202,01 g/mol
Melting Pt: 179…188 °C
Storage Temperature: Ambient
MDL Number: MFCD01927100
CAS Number: 6311-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR