Tantalum pentabromide ≥99.9% (metals basis)

Supplier: Thermo Scientific
Danger

14041.09 14041.30 14041.18
14041.09EA 116 GBP
14041.09 14041.30 14041.18
Tantalum pentabromide ≥99.9% (metals basis)
Tantalum pentabromide

Purity: 99.9% CAS No.: 13451-11-1 Molecular Formula: TaBr5 Formula Weight: 580.49 Physical Form: -8 Mesh Powder Density: 4.67 Appearance: Yellow Odour: Pungent Melting Point: 265° Boiling Point: 348.8° UN No.: 3260 MDL No.: MFCD00049563

Formula: Br₅Ta
MW: 580,47 g/mol
Boiling Pt: 348,8 °C
Melting Pt: 265 °C
Density: 4,67
Storage Temperature: Ambient
MDL Number: MFCD00049563
CAS Number: 13451-11-1
EINECS: 236-618-4
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR