Phenylmercury chloride (63,5% Hg) ≥96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Chlorophenylmercury
37113.09
37113.18
37113.09EA
31.5
GBP
37113.09
37113.18
Phenylmercury chloride (63,5% Hg) ≥96%
Phenylmercury chloride
Purity: 96%
CAS No.: 100-56-1
Molecular Formula: C6H5HgCl
Formula Weight: 313.18
Physical Form: Powder
Appearance: White
Melting Point: 246-250°
Solubility: Soluble in benzene, ether, pyridine. Very slightly soluble in water
Merck Reference: 14,7301
UN No.: 2026
MDL No.: MFCD00000463
Formula:
C₆H₅ClHg MW: 313,15 g/mol Melting Pt: 246…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000463 CAS Number: 100-56-1 EINECS: 202-865-1 UN: 2026 ADR: 6.1,III Merck Index: 14,7301 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...