Zinc selenide ≥99.999% (metals basis), Optical Grade Optical Grade
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Zinc (II) selenide
88298.06
88298.14
88298.06EA
0
GBP
88298.06
88298.14
Zinc selenide ≥99.999% (metals basis), Optical Grade Optical Grade
Zinc selenide
Purity: 99.999%
CAS No.: 1315-09-9
Molecular Formula: ZnSe
Formula Weight: 144.33
Physical Form: 3-6mm Pieces
Density: 5.42
Melting Point:
For vacuum deposition
Formula:
SeZn Melting Pt: >1100 °C Density: 5,42 Storage Temperature: Ambient |
MDL Number:
MFCD00011296 CAS Number: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1,III Merck Index: 14,10156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...