Silver cyanate ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Argentous cyanate

45411.06 45411.14
45411.06EA 37.4 GBP
45411.06 45411.14
Silver cyanate ≥98%
Silver cyanate

Purity: 98% CAS No.: 3315-16-0 Molecular Formula: CAgNO Formula Weight: 149.89 Physical Form: Powder Density: 4.0 Appearance: White to grey Odour: Odourless Beilstein Reference: 3655040 UN No.: 3077 MDL No.: MFCD00011512

Formula: CAgNO
MW: 149,89 g/mol
Density: 4,0
Storage Temperature: Ambient
MDL Number: MFCD00011512
CAS Number: 3315-16-0
EINECS: 222-006-4
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR