Acenaphthene 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,2-Dihydroacenaphthylene
201341000
201340050
201345000
ACRO201341000EA
298
GBP
ACRO201341000
ACRO201340050
ACRO201345000
Acenaphthene 99%
Acenaphthene
Formula:
C₁₂H₁₀ MW: 154,21 g/mol Boiling Pt: 279 °C (1013 hPa) Melting Pt: 90…95 °C Density: 1,06 g/cm³ Flash Pt: 135 °C (open cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003807 CAS Number: 83-32-9 EINECS: 201-469-6 UN: 3077 ADR: 9,III Merck Index: 13,00029 |
Specification Test Results
Appearance | White to yellow to brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 91 to 95 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...