Acenaphthenequinone 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Acenaphthylene-1,2-dione

B23466.14 B23466.06 B23466.22
B23466.14EA 56.7 GBP
B23466.14 B23466.06 B23466.22
Acenaphthenequinone 95%
Acenaphthenequinone

Purity: 95% CAS No.: 82-86-0 Molecular Formula: C12H6O2 Formula Weight: 182.18 Melting Point: 253-262° Beilstein Reference: 879172

Formula: C₁₂H₆O₂
MW: 182.18 g/mol
Boiling Pt: 351
Melting Pt: 253…262 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003805
CAS Number: 82-86-0
EINECS: 201-441-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR