Terbium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Warning
|
ampouled under argon
13672.03
13672.06
13672.03EA
115
GBP
13672.03
13672.06
Terbium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Terbium (III) iodide
Purity: 99.99%
CAS No.: 13813-40-6
Molecular Formula: TbI3
Formula Weight: 539.64
Physical Form: Flake, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00049573
Formula:
TbI₃ MW: 539,64 g/mol Melting Pt: 946 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049573 CAS Number: 13813-40-6 EINECS: 237-472-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...