1-Methylisoquinoline ≥97%

Supplier: Thermo Scientific
Warning

H27029.03 H27029.06
H27029.03EA 33.4 GBP
H27029.03 H27029.06
1-Methylisoquinoline ≥97%
1-Methylisoquinoline

Purity: 97% CAS No.: 1721-93-3 Molecular Formula: C10H9N Formula Weight: 143.19 Flash Point: 110°(230°F) Refractive Index: 1.6140 Melting Point: 10-12° Boiling Point: 126-128°/16mm MDL No.: MFCD00006902

Formula: C₁₀H₉N
MW: 143,19 g/mol
Boiling Pt: 126…128 °C (16 mmHg)
Melting Pt: 10…12 °C
Density: 1,078
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006902
CAS Number: 1721-93-3
EINECS: 217-017-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR