5-Bromoindole 99%

Supplier: Thermo Scientific

Synonyms: 5-Bromo-1H-indole

107060050 107060250 107061000
ACRO107060050EA 51.2 GBP
ACRO107060050 ACRO107060250 ACRO107061000
5-Bromoindole 99%
5-Bromoindole
Formula: C₈H₆BrN
MW: 196,05 g/mol
Melting Pt: 89…92 °C
Storage Temperature: Freezer
MDL Number: MFCD00005670
CAS Number: 10075-50-0
EINECS: 233-208-7

Order Now

Specification Test Results

Appearance White to light brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 89 to 92 °C
GC ≥98.5 %
Pore volume 0.80 to 1.00 ml/g
Carbon (C) ≥16 %
Volatile matter ≤4 % (110 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR