5-Fluoro-1-indanone ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Fluoro-2,3-dihydro-1H-inden-1-one
L10949.03
L10949.06
L10949.03EA
0
GBP
L10949.03
L10949.06
5-Fluoro-1-indanone ≥99%
5-Fluoro-1-indanone
Purity: 99%
CAS No.: 700-84-5
Molecular Formula: C9H7FO
Formula Weight: 150.15
Flash Point: 109°(228°F)
Melting Point: 37-40°
Boiling Point: 92-93°/3mm
Beilstein Reference: 1617799
MDL No.: MFCD00041031
Formula:
C₉H₇FO MW: 150,15 g/mol Boiling Pt: 92…93 °C (3 mmHg) Melting Pt: 37…40 °C Density: 1,35 g/cm³ (20 °C) Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041031 CAS Number: 700-84-5 EINECS: 615-057-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...