(Diethylamino)sulphur trifluoride

Supplier: Molekula

Synonyms: DAST

15439535-5G 15439535-25G
MOLE15439535-5GEA 174 GBP
MOLE15439535-5G MOLE15439535-25G
(Diethylamino)sulphur trifluoride
(Diethylamino)sulphur trifluoride
Formula: C₄H₁₀F₃NS
MW: 161,19 g/mol
Density: 1,316 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000363
CAS Number: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR