Triethyl phosphite 98%
Supplier: Thermo Scientific
|
Warning
|
139691000
139695000
139690000
ACRO139691000EA
48.6
GBP
ACRO139691000
ACRO139695000
139690000.
Triethyl phosphite 98%
Triethyl phosphite
Formula:
C₆H₁₅O₃P MW: 166,16 g/mol Boiling Pt: 156…158 °C (760 mmHg) Melting Pt: –112 °C Density: 0,965 g/cm³ Flash Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009084 CAS Number: 122-52-1 EINECS: 204-552-5 UN: 2323 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4120 to 1.4140 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...