Phenyltrimethoxysilane 85%

Supplier: Thermo Scientific
Warning

Synonyms: Trimethoxy(phenyl)silane

370640500 370642500 370640010
ACRO370640500EA 52.6 GBP
ACRO370640500 ACRO370642500 ACRO370640010
Phenyltrimethoxysilane 85%
Phenyltrimethoxysilane
Formula: C₉H₁₄O₃Si
MW: 198,29 g/mol
Boiling Pt: 233 °C
Melting Pt: –25 °C
Density: 1,062 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00025689
CAS Number: 2996-92-1
EINECS: 221-066-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4730 to 1.4750 (20°C, 589 nm)
GC ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR