Phenyltriethoxysilane ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Phenyltriethoxysilane, Triethoxy(phenyl)silane, Triethoxyphenylsilane
L04684.18
L04684.30
L04684.0B
L04684.18EA
21.3
GBP
L04684.18
L04684.30
L04684.0B
Phenyltriethoxysilane ≥98%
Phenyltriethoxysilane
Purity: 98%
CAS No.: 780-69-8
Molecular Formula: C12H20Si
Formula Weight: 240.38
Flash Point: 96°(204°F)
Refractive Index: 1.4610
Boiling Point: 235°
Beilstein Reference: 2940602
MDL No.: MFCD00009065
Formula:
C₁₂H₂₀O₃Si MW: 240,37 g/mol Boiling Pt: 235 °C Melting Pt: 41 °C Density: 0,996 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009065 CAS Number: 780-69-8 EINECS: 212-305-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...