Triisopropylsilane ≥98%

Supplier: Thermo Scientific
Warning

L09585.14 L09585.06 L09585.22 L09585.PG L09585.PF
L09585.14EA 80.4 GBP
L09585.14 L09585.06 L09585.22 L09585.PG L09585.PF
Triisopropylsilane ≥98%
Triisopropylsilane

Purity: 98% CAS No.: 6485-79-6 Molecular Formula: C9H22Si Formula Weight: 158.36 Flash Point: 35°(95°F) Refractive Index: 1.4340 Odour: Mild Boiling Point: 166° Beilstein Reference: 1733718 UN No.: 1993 MDL No.: MFCD00009657

Formula: C₉H₂₂Si
MW: 158,36 g/mol
Boiling Pt: 166 °C
Density: 0,773
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009657
CAS Number: 6485-79-6
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR