(3-Mercaptopropyl)trimethoxysilane 85% tech.
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Trimethoxy(3-mercaptopropyl)silane, 3-(Trimethoxysilyl)propane-1-thiol, 3-(Trimethoxysilyl)-1-propanethiol
174650500
174652500
ACRO174650500EA
87.7
GBP
ACRO174650500
ACRO174652500
(3-Mercaptopropyl)trimethoxysilane 85% tech.
(3-Mercaptopropyl)trimethoxysilane
Formula:
HSH₂CCH₂CH₂Si(OCH₃)₃ MW: 196.34 g/mol Boiling Pt: 215 Melting Pt: –50 °C Density: 1.05 g/cm³ Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004901 CAS Number: 4420-74-0 EINECS: 224-588-5 UN: 3082 ADR: 9 III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4410 to 1.4450 (20°C, 589 nm) |
GC | ≥80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...