Methyltrimethoxysilane ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Trimethoxy(methyl)silane
B23594.AK
B23594.AU
B23594.AKEA
33.4
GBP
B23594.AK
B23594.AU
Methyltrimethoxysilane ≥97%
Methyltrimethoxysilane
Purity: 97%
CAS No.: 1185-55-3
Molecular Formula: CH3Si(OCH3)3
Formula Weight: 136.23
Flash Point: 9°(48°F)
Refractive Index: 1.3700
Boiling Point: 102-103°
Beilstein Reference: 1736151
UN No.: 1993
MDL No.: MFCD00008342
Formula:
C₄H₁₂O₃Si Boiling Pt: 102…103 °C Melting Pt: 11 °C Density: 0,955 Flash Pt: 9 °C (48 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008342 CAS Number: 1185-55-3 EINECS: 214-685-0 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...