Decyltriethoxysilane ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Triethoxy(decyl)silane

L16965.14 L16965.22
L16965.14EA 89.2 GBP
L16965.14 L16965.22
Decyltriethoxysilane ≥98%
Decyltriethoxysilane

Purity: 98% CAS No.: 2943-73-9 Molecular Formula: C16H36O3Si Formula Weight: 304.55 Flash Point: 110°(230°F) Refractive Index: 1.4220 Boiling Point: 150°/8mm Beilstein Reference: 2413155 MDL No.: MFCD00069132

Formula: C₁₆H₃₆O₃Si
MW: 304,55 g/mol
Boiling Pt: 150 °C (8 mmHg)
Density: 0,879
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00069132
CAS Number: 2943-73-9
EINECS: 220-940-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR