(3-Chloropropyl)trimethoxysilane 98+%
Supplier: Thermo Scientific
Synonyms:
3-(Trimethoxysilyl)propylchloride
, 3-Chloropropyltrimethoxysilane
, Trimethoxy(3-chloropropyl)silane
ACRO137280250EA
63.7
GBP
ACRO137280250
ACRO137281000
ACRO137285000
(3-Chloropropyl)trimethoxysilane 98+%
(3-Chloropropyl)trimethoxysilane
Formula:
ClH₂CCH₂CH₂Si(OCH₃)₃ MW: 198,72 g/mol Boiling Pt: 195…196 °C (750 mmHg) Density: 1,07 g/cm³ Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000997 CAS Number: 2530-87-2 EINECS: 219-787-9 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4210 (20°C, 589 nm) |
GC | ≥98.0 % |
Specific optical rotation | +16° to +20° (20°C, 589 nm) (c=5% in Methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...