Germanium(IV) ethoxide ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate

14679.03 14679.06 14679.14
14679.03EA 48.7 GBP
14679.03 14679.06 14679.14
Germanium(IV) ethoxide ≥97%
Germanium (IV) ethoxide

Purity: 97% CAS No.: 14165-55-0 Molecular Formula: C8H20GeO4 Formula Weight: 252.84 Flash Point: 49°(121°F) Physical Form: Liquid Density: 1.14 Refractive Index: 1.4070 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Alcohol-like Melting Point: -72° Boiling Point: 185.5° Solubility: Soluble in anhydrous alcohol and benzene. Hydrolyzes in water UN No.: 1993 MDL No.: MFCD00015121

Formula: C₈H₂₀GeO₄
MW: 252,85 g/mol
Boiling Pt: 185,5 °C
Melting Pt: –72 °C
Density: 1,14
Flash Pt: 49 °C (121 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015121
CAS Number: 14165-55-0
EINECS: 238-007-8
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR