Silicon(II) oxide, Patinal®
Supplier: Merck
Synonyms:
Silicon monoxide, Silicon oxide
1.07716.0100
1.07716.1000
1.07716.0100EA
0
GBP
1.07716.0100
1.07716.1000
Silicon(II) oxide, Patinal®
Silicon (II) oxide
Formula:
SiO MW: 44,08 g/mol Boiling Pt: 1880 °C (1013 hPa) Melting Pt: 1702 °C Density: 2,13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151536 CAS Number: 10097-28-6 EINECS: 233-232-8 Merck Index: 12,08640 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...