Magnesium fluoride, Patinal®

Supplier: Merck

1.05834.0100 1.05834.1000
1.05834.0100EA 0 GBP
1.05834.0100 1.05834.1000
Magnesium fluoride, Patinal®
Magnesium fluoride
Formula: MgF₂
MW: 62,3 g/mol
Boiling Pt: 2260 °C (1013 hPa)
Melting Pt: 1266 °C
Density: 3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Merck Index: 12,05701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR