Dipotassium hexahydroxoplatinate ≥99.95% (metals basis) Pt ≥ 51.5%, Premion®

Supplier: Thermo Scientific
Warning

Synonyms: Potassium hexahydroxoplatinate, Dipotassium platinate, Potassium hexahydroxyplatinate(IV)

41014.03 41014.06
41014.03EA 165 GBP
41014.03 41014.06
Dipotassium hexahydroxoplatinate ≥99.95% (metals basis) Pt ≥ 51.5%, Premion®
Dipotassium hexahydroxoplatinate

Purity: 99.95% CAS No.: 12285-90-4 Molecular Formula: K2Pt(OH)6 Formula Weight: 375.34 Physical Form: Powder

Formula: H₆K₂O₆Pt
MW: 375,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064671
CAS Number: 12285-90-4
EINECS: 235-554-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR