Potassium periodate ≥99%

Supplier: Thermo Fisher Scientific
Danger

A11308.0E A11308.22 A11308.36
A11308.0EEA 655 GBP
A11308.0E A11308.22 A11308.36
Potassium periodate ≥99%
Potassium periodate

Purity: 99% CAS No.: 7790-21-8 Molecular Formula: KIO4 Formula Weight: 230.00 Melting Point: 582° Merck Reference: 14,7654 MDL No.: MFCD00011407

Formula: KlO₄
Melting Pt: 582 °C
Density: 3,618
Storage Temperature: Ambient
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 1479
ADR: 5.1,II
Merck Index: 14,7654

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR