2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L16563.06
L16563.14
L16563.06EA
27.5
GBP
L16563.06
L16563.14
2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane ≥97%
2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane
Purity: 97%
CAS No.: 1020-84-4
Molecular Formula: C8H28N4Si4
Formula Weight: 292.68
Flash Point: 66°(150°F)
Odour: Ammonia-like
Boiling Point: 224-225°
Beilstein Reference: 976248
UN No.: 2923
MDL No.: MFCD00046123
Formula:
C₈H₂₈N₄Si₄ Boiling Pt: 224…225 °C Melting Pt: 94…98 °C Density: 0,95 g/cm³ (20 °C) Flash Pt: 66 °C (150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046123 CAS Number: 1020-84-4 EINECS: 213-817-4 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...