2,3,4',6-Tetrachlorobiphenyl (PCB No. 64)

Supplier: LGC Standards PROMOCHEM

PROOCIL-PCB-64-CEA 970 GBP
PROOCIL-PCB-64-C
2,3,4',6-Tetrachlorobiphenyl (PCB No. 64)
2,3,4',6-Tetrachlorobiphenyl (PCB No. 64)
Formula: C₁₂H₆Cl₄
MW: 291,99 g/mol
CAS Number: 52663-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR