2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)

Supplier: LGC Standards PROMOCHEM

PROOCIL-PCB-110-CEA 857 GBP
PROOCIL-PCB-110-C
2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)
2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)
Formula: C₁₂H₅Cl₅
MW: 326,44 g/mol
CAS Number: 38380-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR